COM448636562 Combi-Blocks, Inc.

tert-Butyl 8-nitro-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate

  • MDL number MFCD19704739
  • Cas Number : 1415819-76-9
  • Molecular Weight : 362.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0469

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0469
Availability details for
Quantity
  • Availability
  • Add to Cart