COM448636712 Combi-Blocks, Inc.

1-(7-Chloro-6-nitro-3,4-dihydroisoquinolin-2(1h)-yl)-2,2,2-trifluoroethanone

  • MDL number MFCD20921906
  • Cas Number : 1097920-65-4
  • Molecular Weight : 308.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0646
Availability details for
Quantity
  • Availability
  • Add to Cart