COM448636714 Combi-Blocks, Inc.

2-Bromomethyl-6-pyridine carboxylic acid ethyl ester

  • MDL number MFCD09026929
  • Cas Number : 97278-44-9
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0649

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0649
Availability details for
Quantity
  • Availability
  • Add to Cart