COM448636727 Combi-Blocks, Inc.

1-Benzyl-3-iodo-7-azaindole

  • MDL number MFCD09834873
  • Cas Number : 1093759-51-3
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0667

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0667
Availability details for
Quantity
  • Availability
  • Add to Cart