COM448636738 Combi-Blocks, Inc.

2,3-Dihydro-1h-cyclopenta[b]quinoline

  • MDL number MFCD18448962
  • Cas Number : 5661-06-3
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0679

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0679
Availability details for
Quantity
  • Availability
  • Add to Cart