COM448636786 Combi-Blocks, Inc.

4-Chloro-2-(3,5-dichlorophenyl)-5-(1-methylhydrazino)-2,3-dihydropyridazin-3-one

  • MDL number MFCD00052035
  • Cas Number : 175135-84-9
  • Molecular Weight : 319.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0735
Availability details for
Quantity
  • Availability
  • Add to Cart