COM448636787 Combi-Blocks, Inc.

N-(2-Hydroxyethyl)-2-nitrobenzenesulfonamide

  • MDL number MFCD00458943
  • Cas Number : 18226-11-4
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0736

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0736
Availability details for
Quantity
  • Availability
  • Add to Cart