COM448636817 Combi-Blocks, Inc.

N-Benzyloxymethyl-4-nitroimidazole

  • MDL number MFCD00798167
  • Cas Number : 669713-66-0
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0771

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0771
Availability details for
Quantity
  • Availability
  • Add to Cart