COM448636825 Combi-Blocks, Inc.

5-Chloro-2-methylsulfanyl-pyrimidine-4-carboxylic acid methyl ester

  • MDL number MFCD06496440
  • Cas Number : 79686-03-6
  • Molecular Weight : 218.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0781
Availability details for
Quantity
  • Availability
  • Add to Cart