COM448636833 Combi-Blocks, Inc.

(2-Pyrrolidin-1-ylphenyl)methanol

  • MDL number MFCD08690257
  • Cas Number : 73051-88-4
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0790
Availability details for
Quantity
  • Availability
  • Add to Cart