COM448636910 Combi-Blocks, Inc.

tert-Butyl 4-(3-iodo-1h-indazol-6-yl)piperazine-1-carboxylate

  • MDL number MFCD13183044
  • Cas Number : 744219-44-1
  • Molecular Weight : 428.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0878

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0878
Availability details for
Quantity
  • Availability
  • Add to Cart