COM448636951 Combi-Blocks, Inc.

O-Benzyl-d-serine benzyl ester hcl

  • MDL number MFCD09878761
  • Cas Number : 21948-10-7
  • Molecular Weight : 321.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0924
Availability details for
Quantity
  • Availability
  • Add to Cart