COM448636979 Combi-Blocks, Inc.

4-tert-Butyl-n-(6-chloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidin-4-yl)benzenesulfonamide

  • MDL number MFCD09751194
  • Cas Number : 150727-06-3
  • Molecular Weight : 526.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0957

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0957
Availability details for
Quantity
  • Availability
  • Add to Cart