COM448637105 Combi-Blocks, Inc.

N-Carbobenzoxy-d-leucine dicyclohexylammonium salt

  • MDL number MFCD10566885
  • Cas Number : 7662-58-0
  • Molecular Weight : 446.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1110

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1110
Availability details for
Quantity
  • Availability
  • Add to Cart