COM448637131 Combi-Blocks, Inc.

5-(2-Furyl)-4h-1,2,4-triazol-3-amine

  • MDL number MFCD07427745
  • Cas Number : 3663-61-4
  • Molecular Weight : 150.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1141
Availability details for
Quantity
  • Availability
  • Add to Cart