COM448637140 Combi-Blocks, Inc.

Methyl 4-[(5,6,7,8-tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)carbonyl]benzoate

  • MDL number MFCD12828199
  • Cas Number : 153559-45-6
  • Molecular Weight : 364.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1152
Availability details for
Quantity
  • Availability
  • Add to Cart