COM448637153 Combi-Blocks, Inc.

D-Ethylgonendione

  • MDL number MFCD08457890
  • Cas Number : 21800-83-9
  • Molecular Weight : 286.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1171

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1171
Availability details for
Quantity
  • Availability
  • Add to Cart