COM448637189 Combi-Blocks, Inc.

tert-Butyl 3-(allyloxy)-3-vinylpiperidine-1-carboxylate

  • MDL number MFCD11100937
  • Cas Number : 1065075-83-3
  • Molecular Weight : 267.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1211

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1211
Availability details for
Quantity
  • Availability
  • Add to Cart