COM448637217 Combi-Blocks, Inc.

[1-(2-Chloro-pyrimidin-4-yl)-piperidin-4-yl]-carbamic acid tert-butyl ester

  • MDL number MFCD09028512
  • Cas Number : 596817-49-1
  • Molecular Weight : 312.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1248
Availability details for
Quantity
  • Availability
  • Add to Cart