COM448637219 Combi-Blocks, Inc.

1-Amino-4-hydroxyanthraquinone

  • MDL number MFCD00001223
  • Cas Number : 116-85-8
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1250
Availability details for
Quantity
  • Availability
  • Add to Cart