COM448637232 Combi-Blocks, Inc.

S-2-(Cbz-aminomethyl)pyrrolidine-hcl

  • MDL number MFCD09749850
  • Cas Number : 1033245-45-2
  • Molecular Weight : 270.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1272

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1272
Availability details for
Quantity
  • Availability
  • Add to Cart