COM448637234 Combi-Blocks, Inc.

N-Fmoc-4-methanesulfonylamino-l-phenylalanine

  • MDL number MFCD01860660
  • Cas Number : 266999-22-8
  • Molecular Weight : 480.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1276

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1276
Availability details for
Quantity
  • Availability
  • Add to Cart