COM448637257 Combi-Blocks, Inc.

8-Methoxy-5h-chromeno[4,3-d]pyrimidine-2-thiol

  • MDL number MFCD14585394
  • Cas Number : 1206969-45-0
  • Molecular Weight : 246.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1310
Availability details for
Quantity
  • Availability
  • Add to Cart