COM448637261 Combi-Blocks, Inc.

1-[3-(Trifluoromethyl)phenyl]propylamine

  • MDL number MFCD07784289
  • Cas Number : 473732-59-1
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1314

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1314
Availability details for
Quantity
  • Availability
  • Add to Cart