COM448637296 Combi-Blocks, Inc.

5-(2-Chloro-5-(trifluoromethyl)phenoxy)pyrazin-2-amine

  • MDL number MFCD14585379
  • Cas Number : 1206970-21-9
  • Molecular Weight : 289.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1362
Availability details for
Quantity
  • Availability
  • Add to Cart