COM448637434 Combi-Blocks, Inc.

2,3,4,5-Tetrahydro-8-methyl-1h-pyrido[4,3-b]indole

  • MDL number MFCD05663532
  • Cas Number : 64172-41-4
  • Molecular Weight : 186.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1538

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1538
Availability details for
Quantity
  • Availability
  • Add to Cart