COM448637509 Combi-Blocks, Inc.

tert-Butyl 3-amino-2-oxo-1-pyrrolidineacetic acid

  • MDL number MFCD15071464
  • Cas Number : 1260639-73-3
  • Molecular Weight : 214.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1634
Availability details for
Quantity
  • Availability
  • Add to Cart