COM448637593 Combi-Blocks, Inc.

3-Bromo-n-(5-methylisoxazol-3-yl)propanamide

  • MDL number MFCD01763613
  • Cas Number : 349442-99-5
  • Molecular Weight : 233.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1739
Availability details for
Quantity
  • Availability
  • Add to Cart