COM448637754 Combi-Blocks, Inc.

Scopolamine

  • MDL number MFCD05662373
  • Cas Number : 51-34-3
  • Molecular Weight : 303.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1937

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1937
Availability details for
Quantity
  • Availability
  • Add to Cart