COM448637866 Combi-Blocks, Inc.

3,5-Diethoxy-4-(1h-pyrrol-1-yl)benzaldehyde

  • MDL number MFCD14585238
  • Cas Number : 909853-89-0
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2067
Availability details for
Quantity
  • Availability
  • Add to Cart