COM448637913 Combi-Blocks, Inc.

Methyl (+)-(3r)-7-[4-(4-fluorophenyl)-6-isopropyl-2-(n-methyl-n-methanesulfonylamino)pyrimidin-5-yl]-3-hydroxy-5-oxo-(6e)-heptenoate

  • MDL number MFCD12755726
  • Cas Number : 147118-39-6
  • Molecular Weight : 493.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2127

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2127
Availability details for
Quantity
  • Availability
  • Add to Cart