COM448637977 Combi-Blocks, Inc.

N-2-Hydroxybenzyl-l-serine methyl ester

  • MDL number MFCD05865269
  • Cas Number : 1176623-78-1
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2205

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2205
Availability details for
Quantity
  • Availability
  • Add to Cart