COM448638001 Combi-Blocks, Inc.

4-(2-Chloropyrimidin-4-yl)-n,n-dimethylaniline

  • MDL number MFCD14585486
  • Cas Number : 936939-36-5
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2232
Availability details for
Quantity
  • Availability
  • Add to Cart