COM448638094 Combi-Blocks, Inc.

4-(6-Chloropyridin-2-yl)morpholine

  • MDL number MFCD11848425
  • Cas Number : 330682-30-9
  • Molecular Weight : 198.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2348

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2348
Availability details for
Quantity
  • Availability
  • Add to Cart