COM448638168 Combi-Blocks, Inc.

N-[5-(2-Bromo-acetyl)-2-hydroxy-phenyl]-methanesulfonamide

  • MDL number MFCD06796626
  • Cas Number : 14347-24-1
  • Molecular Weight : 308.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2444

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2444
Availability details for
Quantity
  • Availability
  • Add to Cart