COM448638186 Combi-Blocks, Inc.

Ethyl 5-(chloromethyl)-1,2,4-oxadiazole-3-carboxylate

  • MDL number MFCD11052346
  • Cas Number : 1009620-97-6
  • Molecular Weight : 190.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2465
Availability details for
Quantity
  • Availability
  • Add to Cart