COM448638258 Combi-Blocks, Inc.

1-(4-Bromo-2-methylphenyl)hydrazine, HCl

  • MDL number MFCD04966813
  • Cas Number : 56056-25-8
  • Molecular Weight : 237.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2552
Availability details for
Quantity
  • Availability
  • Add to Cart