COM448638505 Combi-Blocks, Inc.

(R)-3-Amino-3-phenylpropanoic acid ethyl ester HCl

  • MDL number MFCD02259750
  • Cas Number : 340188-50-3
  • Molecular Weight : 229.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2850

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-2850
Availability details for
Quantity
  • Availability
  • Add to Cart