COM448638646 Combi-Blocks, Inc.

N-Acetyl-2-piperidone

  • MDL number MFCD09800363
  • Cas Number : 3326-13-4
  • Molecular Weight : 141.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3014

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3014
Availability details for
Quantity
  • Availability
  • Add to Cart