COM448638745 Combi-Blocks, Inc.

5-(2-Chlorophenyl)-1h-pyrazole-3-carboxylic acid

  • MDL number MFCD05170018
  • Cas Number : 890621-13-3
  • Molecular Weight : 222.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3135
Availability details for
Quantity
  • Availability
  • Add to Cart