COM448639055 Combi-Blocks, Inc.

3,5,6-Trichlorosalicylic acid

  • MDL number MFCD00075245
  • Cas Number : 40932-60-3
  • Molecular Weight : 241.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3556

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3556
Availability details for
Quantity
  • Availability
  • Add to Cart