COM448639194 Combi-Blocks, Inc.

1-Butanesulfonyl chloride

  • MDL number MFCD00007464
  • Cas Number : 2386-60-9
  • Molecular Weight : 156.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3731

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3731
Availability details for
Quantity
  • Availability
  • Add to Cart