COM448639383 Combi-Blocks, Inc.

P-Aminoethylbenzaldehyde hcl

  • MDL number MFCD09878751
  • Cas Number : 64353-37-3
  • Molecular Weight : 185.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3972

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-3972
Availability details for
Quantity
  • Availability
  • Add to Cart