COM448639602 Combi-Blocks, Inc.

tert-Butyl 7-phenyl-1,4-diazepane-1-carboxylate

  • MDL number MFCD18837826
  • Cas Number : 220898-23-7
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4250
Availability details for
Quantity
  • Availability
  • Add to Cart