COM448639760 Combi-Blocks, Inc.

N-Hydroxy-1-oxy-nicotinamidine

  • MDL number MFCD00208778
  • Cas Number : 92757-16-9
  • Molecular Weight : 153.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4445

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4445
Availability details for
Quantity
  • Availability
  • Add to Cart