COM448639795 Combi-Blocks, Inc.

Pivalaldehyde oxime

  • MDL number MFCD00160762
  • Cas Number : 637-91-2
  • Molecular Weight : 101.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4501

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4501
Availability details for
Quantity
  • Availability
  • Add to Cart