COM448639827 Combi-Blocks, Inc.

Tilmicosin

  • MDL number MFCD00864842
  • Cas Number : 108050-54-0
  • Molecular Weight : 869.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4539

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4539
Availability details for
Quantity
  • Availability
  • Add to Cart