COM448639832 Combi-Blocks, Inc.

2-Methyl-4h-furo[3,2-b]pyrrole-5-carboxylic acid

  • MDL number MFCD07403678
  • Cas Number : 80709-80-4
  • Molecular Weight : 165.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4544
Availability details for
Quantity
  • Availability
  • Add to Cart