COM448639983 Combi-Blocks, Inc.

N1-(2-(2-Chlorophenylthio)benzyl)-n2,n2-dimethylethane-1,2-diamine

  • MDL number MFCD09788617
  • Cas Number : 951624-18-3
  • Molecular Weight : 320.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4738

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4738
Availability details for
Quantity
  • Availability
  • Add to Cart