COM448640044 Combi-Blocks, Inc.

tert-Butyl 2-cyanothiomorpholine-4-carboxylate

  • MDL number MFCD11975781
  • Cas Number : 1311254-50-8
  • Molecular Weight : 228.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4811
Availability details for
Quantity
  • Availability
  • Add to Cart